4-Fluoroaniline Hydroiodide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
F1273-1G
F1273-5G
TCF1273-1GEA
61
USD
TCF1273-1G
TCF1273-5G
4-Fluoroaniline Hydroiodide ≥98.0% (by HPLC, titration analysis)
4-Fluoroaniline Hydroiodide
Formula:
C₆H₆FN·HI |
CAS Number:
85734-19-6 |
Specification Test Results
Appearance | White to Gray to Red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...