4-Fluoroaniline hydrobromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Fluorophenylammonium bromide, 4-Fluoroanilinium bromide, 4-Fluorophenylamine hydrobromide

F1272-25G F1272-5G
TCF1272-25GEA 379 USD
TCF1272-25G TCF1272-5G
4-Fluoroaniline hydrobromide ≥98.0% (by HPLC, titration analysis)
4-Fluoroaniline hydrobromide
Formula: C₆H₆FN·HBr
MW: 192.03 g/mol
CAS Number: 85734-18-5

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR