4,4'-Di(1H-imidazol-1-yl)-1,1'-biphenyl ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 1,1'-(4,4'-Biphenyldiyl)bis(1H-imidazole)

D5777-1G D5777-5G
TCD5777-1GEA 176 USD
TCD5777-1G TCD5777-5G
4,4'-Di(1H-imidazol-1-yl)-1,1'-biphenyl ≥98.0% (by HPLC, total nitrogen)
4,4'-Di(1H-imidazol-1-yl)-1,1'-biphenyl
Formula: C₁₈H₁₄N₄
MW: 286.34 g/mol
MDL Number: MFCD23103195
CAS Number: 855766-92-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR