(Ferrocenylmethyl)trimethylammonium chloride ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: (Trimethylammoniomethyl)ferrocene chloride, FcNCl

F1231-1G F1231-5G
TCF1231-1GEA 86 USD
TCF1231-1G TCF1231-5G
(Ferrocenylmethyl)trimethylammonium chloride ≥98.0% (by HPLC)
(Ferrocenylmethyl)trimethylammonium chloride
Formula: C₁₄H₂₀ClFeN
MW: 293.62 g/mol
CAS Number: 83617-79-2

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 95.0 %
Water max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR