3-(Methoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Bicyclo[1.1.1]pentane-1,3-dicarboxylic acid 1-Monomethyl ester

M3149-200MG M3149-1G
TCM3149-200MGEA 162 USD
TCM3149-200MG TCM3149-1G
3-(Methoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid ≥97.0% (by titrimetric analysis)
3-(Methoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid
Formula: C₈H₁₀O₄
MW: 170.16 g/mol
MDL Number: MFCD20638314
CAS Number: 83249-10-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 138.0 to 142.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR