4-Fluoro-2-formylphenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Fluoro-2-formylbenzeneboronic acid, (4-Fluoro-2-formylphenyl)boronic acid
F1260-1G
F1260-5G
TCF1260-1GEA
107
USD
TCF1260-1G
TCF1260-5G
4-Fluoro-2-formylphenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)
4-Fluoro-2-formylphenylboronic acid
Formula:
C₇H₆BFO₃ MW: 167.93 g/mol Melting Pt: 125 °C |
MDL Number:
MFCD10697426 CAS Number: 825644-26-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | 98.0 to 112.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...