Fluroxypyr 1-Methylheptyl ester ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 1-Methylheptyl [(4-Amino-3,5-dichloro-6-fluoropyridin-2-yl)oxy]acetate, Fluroxypyr Meptyl

F1213-25G F1213-5G
TCF1213-25GEA 55 USD
TCF1213-25G TCF1213-5G
Fluroxypyr 1-Methylheptyl ester ≥98.0% (by HPLC, total nitrogen)
Fluroxypyr 1-Methylheptyl ester
Formula: C₁₅H₂₁Cl₂FN₂O₃
MW: 367.24 g/mol
MDL Number: MFCD00144318
CAS Number: 81406-37-3
EINECS: 279-752-9
UN: 3077
ADR: 9,III
Merck Index: 14,04203

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 56.0 to 60.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR