2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Perfluorobutyl-1,2-epoxypropane

N1041-100G N1041-25G N1041-5G
TCN1041-100GEA 212 USD
TCN1041-100G TCN1041-25G TCN1041-5G
2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane ≥98.0% (by GC)
2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane
Formula: C₇H₅F₉O
MW: 276.10 g/mol
Boiling Pt: 132 °C
MDL Number: MFCD00236085
CAS Number: 81190-28-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR