1,2-Di-tert-butoxy-1,1,2,2-tetramethyldisilane ≥97.0% (by GC)

Supplier: TCI America
Warning

D5736-25G D5736-5G
TCD5736-25GEA 429 USD
TCD5736-25G TCD5736-5G
1,2-Di-tert-butoxy-1,1,2,2-tetramethyldisilane ≥97.0% (by GC)
1,2-Di-tert-butoxy-1,1,2,2-tetramethyldisilane
Formula: C₁₂H₃₀O₂Si₂
MW: 262.54 g/mol
Boiling Pt: 127 °C
Density: 0.84
CAS Number: 78669-53-1

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR