Phenylpiracetam ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2-(2-Oxo-4-phenyl-1-pyrrolidinyl)acetamide, Karfedon, Carphedon

P2604-1G P2604-5G
TCP2604-1GEA 104 USD
TCP2604-1G TCP2604-5G
Phenylpiracetam ≥98.0% (by HPLC, total nitrogen)
Phenylpiracetam
Formula: C₁₂H₁₄N₂O₂
MW: 218.26 g/mol
MDL Number: MFCD01456750
CAS Number: 77472-70-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 130.0 to 134.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR