Dichloro(methyl)(2-phenylethyl)silane ≥98.0% (by GC)

Supplier: TCI America
Danger

D5569-1G D5569-5G
TCD5569-1GEA 84 USD
TCD5569-1G TCD5569-5G
Dichloro(methyl)(2-phenylethyl)silane ≥98.0% (by GC)
Dichloro(methyl)(2-phenylethyl)silane
Formula: C₉H₁₂Cl₂Si
Boiling Pt: 245 °C
Flash Pt: 105 °C
MDL Number: MFCD00018084
CAS Number: 772-65-6
EINECS: 212-253-6
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR