2-Ethynyl-1,3,5-trimethylbenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Ethynyl-2,4,6-trimethylbenzene, 2-Ethynylmesitylene, 2,4,6-Trimethylphenylacetylene

E1361-1G E1361-5G
TCE1361-1GEA 87 USD
TCE1361-1G TCE1361-5G
2-Ethynyl-1,3,5-trimethylbenzene ≥98.0% (by GC)
2-Ethynyl-1,3,5-trimethylbenzene
Formula: C₁₁H₁₂
MW: 144.22 g/mol
Boiling Pt: 215 °C
Melting Pt: 4 °C
Flash Pt: 83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07777099
CAS Number: 769-26-6

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR