Ethyl 2-(trans-4-Aminocyclohexyl)acetate hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: trans-2-(4-Aminocyclohexyl)acetic acid Ethyl ester hydrochloride, Ethyl 2-(trans-4-aminocyclohexyl)acetate hydrochloride

E1374-1G E1374-5G
TCE1374-1GEA 154 USD
TCE1374-1G TCE1374-5G
Ethyl 2-(trans-4-Aminocyclohexyl)acetate hydrochloride ≥98.0% (by titrimetric analysis)
Ethyl 2-(trans-4-Aminocyclohexyl)acetate hydrochloride
Formula: C₁₀H₁₉NO₂·HCl
MW: 221.73 g/mol
MDL Number: MFCD16660366
CAS Number: 76308-26-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR