N-Lauroylglycine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

L0350-1G L0350-5G
TCL0350-1GEA 114 USD
TCL0350-1G TCL0350-5G
N-Lauroylglycine ≥98.0% (by HPLC, titration analysis)
N-Lauroylglycine
Formula: C₁₄H₂₇NO₃
MDL Number: MFCD00153470
CAS Number: 7596-88-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR