Dimethyl 2-(4-Methoxybenzylidene)malonate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-(4-Methoxybenzylidene)malonic acid Dimethyl ester

D5642-25G D5642-5G
TCD5642-25GEA 53 USD
TCD5642-25G TCD5642-5G
Dimethyl 2-(4-Methoxybenzylidene)malonate ≥98.0% (by GC)
Dimethyl 2-(4-Methoxybenzylidene)malonate
Formula: C₁₃H₁₄O₅
MW: 250.25 g/mol
Boiling Pt: 130 °C
MDL Number: MFCD09753105
CAS Number: 7443-25-6
EINECS: 231-185-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR