Dibenzo[b,d]thiophen-2-amine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Aminodibenzothiophene

D5521-1G D5521-200MG
TCD5521-1GEA 252 USD
TCD5521-1G TCD5521-200MG
Dibenzo[b,d]thiophen-2-amine ≥98.0% (by GC, titration analysis)
Dibenzo[b,d]thiophen-2-amine
Formula: C₁₂H₉NS
MW: 199.27 g/mol
MDL Number: MFCD00995227
CAS Number: 7428-91-3

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 130.0 to 134.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR