Hexylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

Synonyms: Hexan-1-amine hydrobromide, 1-Aminohexane hydrobromide, Hexylammonium bromide

H1678-1G H1678-5G
TCH1678-1GEA 45 USD
TCH1678-1G TCH1678-5G
Hexylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
Hexylamine hydrobromide
Formula: C₆H₁₅N·HBr
MW: 182.11 g/mol
CAS Number: 7334-95-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Water max. 1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR