Ethyl 2-[(Ethoxycarbonothioyl)thio]propionate ≥95.0% (by GC)

Supplier: TCI America

E1403-1G E1403-5G
TCE1403-1GEA 241 USD
TCE1403-1G TCE1403-5G
Ethyl 2-[(Ethoxycarbonothioyl)thio]propionate ≥95.0% (by GC)
Ethyl 2-[(Ethoxycarbonothioyl)thio]propionate
Formula: C₈H₁₄O₃S₂
Storage Temperature: Refrigerator
MDL Number: MFCD30474932
CAS Number: 73232-07-2

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 95.0 %
Specific gravity (20/20) 1.1240 to 1.1340


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR