Propionaldehyde 2,4-dinitrophenylhydrazone ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
P1166-10G
P1166-1G
TCP1166-10GEA
206
USD
TCP1166-10G
TCP1166-1G
Propionaldehyde 2,4-dinitrophenylhydrazone ≥97.0% (by HPLC, titration analysis)
Propionaldehyde 2,4-dinitrophenylhydrazone
Formula:
C₉H₁₀N₄O₄ MW: 238.20 g/mol |
MDL Number:
MFCD00191484 CAS Number: 725-00-8 UN: 1325 ADR: 4.1,III |
Specification Test Results
Appearance | Orange to Brown powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 148.0 to 155.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...