Fomesafen ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-[2-Chloro-4-(trifluoromethyl)phenoxy]-N-(methylsulphonyl)-2-nitrobenzamide

F1236-1G F1236-25G
TCF1236-1GEA 37 USD
TCF1236-1G TCF1236-25G
Fomesafen ≥98.0% (by HPLC, titration analysis)
Fomesafen
Formula: C₁₅H₁₀ClF₃N₂O₆S
MW: 438.76 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01632756
CAS Number: 72178-02-0
EINECS: 276-439-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 220.0 to 224.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR