Ethyl (3-Hydroxyphenyl)carbamate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: (3-Hydroxyphenyl)carbamic acid Ethyl ester

E1227-1G E1227-5G
TCE1227-1GEA 101 USD
TCE1227-1G TCE1227-5G
Ethyl (3-Hydroxyphenyl)carbamate ≥97.0% (by GC)
Ethyl (3-Hydroxyphenyl)carbamate
Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
MDL Number: MFCD00463773
CAS Number: 7159-96-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR