10-Phenylphenothiazine ≥98.0% (by GC)

Supplier: TCI America

P2470-1G P2470-5G
TCP2470-1GEA 145 USD
TCP2470-1G TCP2470-5G
10-Phenylphenothiazine ≥98.0% (by GC)
10-Phenylphenothiazine
Formula: C₁₈H₁₃NS
Boiling Pt: 223 °C
MDL Number: MFCD00156238
CAS Number: 7152-42-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR