Dimethyl 2,2'-Oxydiacetate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Diglycolic acid Dimethyl ester, 2,2'-Oxydiacetic acid Dimethyl ester, Dimethyl Diglycolate

D5721-1G D5721-5G
TCD5721-1GEA 105 USD
TCD5721-1G TCD5721-5G
Dimethyl 2,2'-Oxydiacetate ≥98.0% (by GC)
Dimethyl 2,2'-Oxydiacetate
Formula: C₆H₁₀O₅
Boiling Pt: 126 °C
MDL Number: MFCD01026217
CAS Number: 7040-23-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR