[Oxalylbis(azanediyl)]bis(ethane-2,1-diyl) bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate] ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
Antioxidant MD-697, (Oxalylbis(azanediyl))bis(ethane-2,1-diyl) bis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate)
O0526-25G
O0526-5G
TCO0526-25GEA
267
USD
TCO0526-25G
TCO0526-5G
[Oxalylbis(azanediyl)]bis(ethane-2,1-diyl) bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate] ≥98.0% (by HPLC, total nitrogen)
[Oxalylbis(azanediyl)]bis(ethane-2,1-diyl) bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate]
Formula:
C₄₀H₆₀N₂O₈ MW: 696.93 g/mol |
MDL Number:
MFCD01940880 CAS Number: 70331-94-1 EINECS: 274-572-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 178.0 to 182.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...