Ethyl 2-(4-Cyanophenyl)hydrazinecarboxylate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-(4-Cyanophenyl)hydrazinecarboxylic acid Ethyl ester, 1-(4-Cyanophenyl)-2-(ethoxycarbonyl)hydrazine

E1299-1G E1299-5G
TCE1299-1GEA 55 USD
TCE1299-1G TCE1299-5G
Ethyl 2-(4-Cyanophenyl)hydrazinecarboxylate ≥98.0% (by GC)
Ethyl 2-(4-Cyanophenyl)hydrazinecarboxylate
Formula: C₁₀H₁₁N₃O₂
MW: 205.22 g/mol
CAS Number: 700371-70-6

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 135.0 to 139.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR