2,4-Dimethyl-3-cyclohexene-1-carboxaldehyde (mixture of isomers) ≥98.0% (by GC)

Supplier: TCI America

D5743-100G D5743-25G
TCD5743-100GEA 109 USD
TCD5743-100G TCD5743-25G
2,4-Dimethyl-3-cyclohexene-1-carboxaldehyde (mixture of isomers) ≥98.0% (by GC)
2,4-Dimethyl-3-cyclohexene-1-carboxaldehyde (mixture of isomers)
Formula: C₉H₁₄O
Boiling Pt: 189 °C
MDL Number: MFCD00221550
CAS Number: 68039-49-6
EINECS: 268-264-1

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR