Hexadecyl 3,5-Di-tert-butyl-4-hydroxybenzoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Hexadecyl 3,5-di-tert-butyl-4-hydroxybenzoate, 3,5-Di-tert-butyl-4-hydroxybenzoic acid Hexadecyl ester

H1709-25G H1709-5G
TCH1709-25GEA 53 USD
TCH1709-25G TCH1709-5G
Hexadecyl 3,5-Di-tert-butyl-4-hydroxybenzoate ≥98.0% (by GC)
Hexadecyl 3,5-Di-tert-butyl-4-hydroxybenzoate
Formula: C₃₁H₅₄O₃
MW: 474.77 g/mol
MDL Number: MFCD00180757
CAS Number: 67845-93-6
EINECS: 267-342-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR