4-Ethoxyphenyl trans-4-Propylcyclohexanecarboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Trans-4-ethoxyphenyl 4-propylcyclohexanecarboxylate, trans-4-Propylcyclohexanecarboxylic acid 4-Ethoxyphenyl ester

E1395-100G E1395-25G
TCE1395-100GEA 388 USD
TCE1395-100G TCE1395-25G
4-Ethoxyphenyl trans-4-Propylcyclohexanecarboxylate ≥98.0% (by GC)
4-Ethoxyphenyl trans-4-Propylcyclohexanecarboxylate
Formula: C₁₈H₂₆O₃
MW: 290.40 g/mol
MDL Number: MFCD11053417
CAS Number: 67589-39-3
EINECS: 266-744-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) max. 98.0 %
Mesomorphic range 33.0 to 78.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR