2,4,6-Tri-tert-butylpyrimidine ≥98.0% (by GC)

Supplier: TCI America
Warning

T3693-1G T3693-5G
TCT3693-1GEA 65 USD
TCT3693-1G TCT3693-5G
2,4,6-Tri-tert-butylpyrimidine ≥98.0% (by GC)
2,4,6-Tri-tert-butylpyrimidine
Formula: C₁₆H₂₈N₂
MDL Number: MFCD03094045
CAS Number: 67490-21-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR