(5-Ethyl-1,3-dioxan-5-yl)methyl acrylate ≥98.0% (by GC) stabilized

Supplier: TCI America
Danger


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

E1334-100G E1334-500G
TCE1334-100GEA 134.33 USD
TCE1334-100G TCE1334-500G
(5-Ethyl-1,3-dioxan-5-yl)methyl acrylate ≥98.0% (by GC) stabilized
(5-Ethyl-1,3-dioxan-5-yl)methyl acrylate
Formula: C₁₀H₁₆O₄
Boiling Pt: 78 °C
MDL Number: MFCD28046653
CAS Number: 66492-51-1
EINECS: 266-380-7
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 2.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR