4-Fluorobenzylamine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

F1255-1G F1255-5G
TCF1255-1GEA 41 USD
TCF1255-1G TCF1255-5G
4-Fluorobenzylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Fluorobenzylamine hydrochloride
Formula: C₇H₈FN·HCl
MDL Number: MFCD00012861
CAS Number: 659-41-6
EINECS: 211-534-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR