Nonamethylene Glycol Dimethacrylate ≥98.0% (by GC) stabilized

Supplier: TCI America
Warning

Synonyms: 1,9-Nonanedioldimethacrylate, 1,9-Nonanediol Dimethacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

N1089-25G N1089-500G
TCN1089-25GEA 30.08 USD
TCN1089-25G TCN1089-500G
Nonamethylene Glycol Dimethacrylate ≥98.0% (by GC) stabilized
Nonamethylene Glycol Dimethacrylate
Formula: C₁₇H₂₈O₄
MW: 296.41 g/mol
MDL Number: MFCD00080593
CAS Number: 65833-30-9

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR