(3,4-Epoxycyclohexyl)methyl acrylate ≥94.0% (by GC) stabilized

Supplier: TCI America


Stabilization: Stabilised with hydroquinone (HQ)

E1384-5G E1384-25G
TCE1384-5GEA 41 USD
TCE1384-5G TCE1384-25G
(3,4-Epoxycyclohexyl)methyl acrylate ≥94.0% (by GC) stabilized
(3,4-Epoxycyclohexyl)methyl acrylate
Formula: C₁₀H₁₄O₃
Boiling Pt: 105 °C
MDL Number: MFCD28364662
CAS Number: 64630-63-3

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR