1-(Furan-2-yl)but-3-en-1-ol ≥95.0% (by GC)

Supplier: TCI America

Synonyms: alpha-2-Propen-1-yl-2-furanmethanol

F1226-1G F1226-5G
TCF1226-1GEA 47 USD
TCF1226-1G TCF1226-5G
1-(Furan-2-yl)but-3-en-1-ol ≥95.0% (by GC)
1-(Furan-2-yl)but-3-en-1-ol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
MDL Number: MFCD03093935
CAS Number: 6398-51-2

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %
Specific gravity (20/20) 1.0420 to 1.0520

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR