Desonide ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: (11beta,16alpha)-11,21-Dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, Budesonide Acetonide

D5578-1G D5578-250MG D5578-5G
TCD5578-1GEA 72 USD
TCD5578-1G TCD5578-250MG TCD5578-5G
Desonide ≥98.0% (by HPLC)
Desonide
Formula: C₂₄H₃₂O₆
MW: 416.51 g/mol
Melting Pt: 275 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00866151
CAS Number: 638-94-8
EINECS: 211-351-6
Merck Index: 14,02928

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +130 to +134 deg (C=0.5, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR