2-(4-Hydroxyphenyl)benzo[b]thiophene-6-ol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-(4-Hydroxyphenyl)benzo[b]thiophen-6-ol

H1628-200MG H1628-50MG
TCH1628-200MGEA 347 USD
TCH1628-200MG TCH1628-50MG
2-(4-Hydroxyphenyl)benzo[b]thiophene-6-ol ≥98.0% (by GC)
2-(4-Hydroxyphenyl)benzo[b]thiophene-6-ol
Formula: C₁₄H₁₀O₂S
MW: 242.29 g/mol
MDL Number: MFCD16619490
CAS Number: 63676-22-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR