2,5-Dibromoterephthalaldehyde ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
2,5-Dibromobenzene-1,4-dicarbaldehyde, 2,5-Dibromo-1,4-diformylbenzene, 2,5-Dibromo-1,4-benzenedicarboxaldehyde
D5793-1G
D5793-250MG
TCD5793-1GEA
292
USD
TCD5793-1G
TCD5793-250MG
2,5-Dibromoterephthalaldehyde ≥97.0% (by GC)
2,5-Dibromoterephthalaldehyde
Formula:
C₈H₄Br₂O₂ MW: 291.93 g/mol |
MDL Number:
MFCD11521314 CAS Number: 63525-48-4 |
Specification Test Results
Appearance | White to Green to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 189.0 to 193.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...