Propoxybenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Phenyl propyl ether

P2607-25ML P2607-5ML
TCP2607-25MLEA 395 USD
TCP2607-25ML TCP2607-5ML
Propoxybenzene ≥98.0% (by GC)
Propoxybenzene
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 190 °C
Melting Pt: –28 °C
MDL Number: MFCD00039935
CAS Number: 622-85-5
EINECS: 210-756-5

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR