Di(N-succinimidyl) L-Tartrate
Supplier: TCI America
Synonyms:
(2R,3R)-2,3-Dihydroxysuccinic acid Di(N-succinimidyl) ester, L-Tartaric acid Di(N-succinimidyl) ester, Di(N-succinimidyl) (2R,3R)-2,3-Dihydroxysuccinate
D5500-250MG
D5500-50MG
TCD5500-250MGEA
276
USD
TCD5500-250MG
TCD5500-50MG
Di(N-succinimidyl) L-Tartrate
Di(N-succinimidyl) L-Tartrate
Formula:
C₁₂H₁₂N₂O₁₀ |
CAS Number:
62069-75-4 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Elemental analysis (Nitrogen) | 7.70 to 8.50 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...