(-)-Synephrine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (R)-(-)-4-[1-Hydroxy-2-(methylamino)ethyl]phenol, (R)-(-)-4-Hydroxy-alpha-(methylaminomethyl)benzyl alcohol, (R)-(-)-1-(4-Hydroxyphenyl)-2-(methylamino)ethanol

Y0012-1G Y0012-5G
TCY0012-1GEA 142 USD
TCY0012-1G TCY0012-5G
(-)-Synephrine ≥98.0% (by HPLC, titration analysis)
(-)-Synephrine
Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
Storage Temperature: Refrigerator
CAS Number: 614-35-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -58.0 to -63.0 deg (C=2, 1mol/L HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR