2-Hydroxyanthracene ≥93.0% (by GC)

Supplier: TCI America

H1565-1G H1565-200MG
TCH1565-1GEA 420 USD
TCH1565-1G TCH1565-200MG
2-Hydroxyanthracene ≥93.0% (by GC)
2-Hydroxyanthracene
Formula: C₁₄H₁₀O
MDL Number: MFCD06653605
CAS Number: 613-14-9

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystaline
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR