Ethynyl(diphenyl)phosphine oxide ≥98.0% (by GC)

Supplier: TCI America
Warning

E1310-1G E1310-200MG
TCE1310-1GEA 303 USD
TCE1310-1G TCE1310-200MG
Ethynyl(diphenyl)phosphine oxide ≥98.0% (by GC)
Ethynyl(diphenyl)phosphine oxide
Formula: C₁₄H₁₁OP
MW: 226.21 g/mol
CAS Number: 6104-48-9

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR