Olsalazine Sodium ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Mordant Yellow 5, Disodium 5,5'-(Diazene-1,2-diyl)bis(2-hydroxybenzoate), 5,5'-Azobis(2-hydroxybenzoic acid sodium salt), 4,4'-Dihydroxyazobenzene-3,3'-dicarboxylic acid disodium salt

O0517-1G O0517-250MG
TCO0517-1GEA 172 USD
TCO0517-1G TCO0517-250MG
Olsalazine Sodium ≥98.0% (by HPLC)
Olsalazine Sodium
Formula: C₁₄H₈N₂Na₂O₆
MDL Number: MFCD00013300
CAS Number: 6054-98-4
EINECS: 227-975-7
Merck Index: 14,06841

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR