4-Methylthiophene-2-carboxaldehyde ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 2-Formyl-4-methylthiophene, 4-Methylthiophene-2-carbaldehyde

M3054-1G M3054-5G
TCM3054-1GEA 78 USD
TCM3054-1G TCM3054-5G
4-Methylthiophene-2-carboxaldehyde ≥95.0% (by GC)
4-Methylthiophene-2-carboxaldehyde
Formula: C₆H₆OS
Boiling Pt: 86 °C
Flash Pt: 92 °C
MDL Number: MFCD00159561
CAS Number: 6030-36-0

Order Now

Specification Test Results

Appearance Colourless to Red to Green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR