6-Methoxy-2-naphthalenemethanol ≥98.0% (by GC)

Supplier: TCI America

M2931-1G M2931-5G
TCM2931-1GEA 83 USD
TCM2931-1G TCM2931-5G
6-Methoxy-2-naphthalenemethanol ≥98.0% (by GC)
6-Methoxy-2-naphthalenemethanol
Formula: C₁₂H₁₂O₂
MDL Number: MFCD06202657
CAS Number: 60201-22-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR