2,4,4-Trimethylpentan-2-amine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T3784-1G T3784-5G
TCT3784-1GEA 30 USD
TCT3784-1G TCT3784-5G
2,4,4-Trimethylpentan-2-amine hydrochloride ≥98.0% (by titrimetric analysis)
2,4,4-Trimethylpentan-2-amine hydrochloride
Formula: C₈H₁₉N·HCl
MDL Number: MFCD30182014
CAS Number: 58618-91-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR