Ethyl 2-(4-Oxocyclohexyl)acetate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl 2-(4-oxocyclohexyl)acetate, 2-(4-Oxocyclohexyl)acetic acid Ethyl ester

E1375-1G E1375-5G
TCE1375-1GEA 62 USD
TCE1375-1G TCE1375-5G
Ethyl 2-(4-Oxocyclohexyl)acetate ≥95.0% (by GC)
Ethyl 2-(4-Oxocyclohexyl)acetate
Formula: C₁₀H₁₆O₃
Boiling Pt: 96 °C
MDL Number: MFCD11045783
CAS Number: 58012-34-3

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR