2'-Methoxy-1'-acetonaphthone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Acetyl-2-methoxynaphthalene, 1-(2-Methoxynaphthalen-1-yl)ethanone

M2875-1G M2875-5G
TCM2875-1GEA 119 USD
TCM2875-1G TCM2875-5G
2'-Methoxy-1'-acetonaphthone ≥98.0% (by GC)
2'-Methoxy-1'-acetonaphthone
Formula: C₁₃H₁₂O₂
MW: 200.24 g/mol
Boiling Pt: 158 °C
MDL Number: MFCD11553477
CAS Number: 5672-94-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR