1,3,5-Tris(trimethylsilyl)benzene ≥95.0% (by GC)

Supplier: TCI America

T3526-1G T3526-5G
TCT3526-1GEA 110 USD
TCT3526-1G TCT3526-5G
1,3,5-Tris(trimethylsilyl)benzene ≥95.0% (by GC)
1,3,5-Tris(trimethylsilyl)benzene
Formula: C₁₅H₃₀Si₃
MW: 294.66 g/mol
Boiling Pt: 244 °C
MDL Number: MFCD00093247
CAS Number: 5624-60-2

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR