Triisocyanato(methyl)silane ≥96.0% (by total nitrogen basis)

Supplier: TCI America
Danger

T3683-25G T3683-5G
TCT3683-25GEA 201 USD
TCT3683-25G TCT3683-5G
Triisocyanato(methyl)silane ≥96.0% (by total nitrogen basis)
Triisocyanato(methyl)silane
Formula: C₄H₃N₃O₃Si
MW: 169.17 g/mol
Boiling Pt: 171 °C
Melting Pt: 3 °C
MDL Number: MFCD00061499
CAS Number: 5587-61-1
UN: 2478
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (with Total Nitrogen) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR